Ethyl picolinoylacetate 95%

Supplier: Thermo Scientific

Synonyms: Ethyl 3-oxo-3-(2-pyridyl)propionate, Ethyl 3-oxo-3-(pyridin-2-yl)propanoate

425470025 425470100
ACRO425470025EA 110 EUR
ACRO425470025 ACRO425470100
Ethyl picolinoylacetate 95%
Ethyl picolinoylacetate
Formula: C₁₀H₁₁NO₃
MW: 193,20 g/mol
Boiling Pt: 100…104 °C (,3 mmHg)
Density: 1,164 g/cm³
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00094022
CAS Number: 26510-52-1

Order Now

Specification Test Results

Appearance Clear straw yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5190 (20°C, 589 nm)
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR