3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%
Supplier: Thermo Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid
L19803.03
L19803.MD
L19803.03EA
168
EUR
L19803.03
L19803.MD
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
Purity: 97%
CAS No.: 284493-65-8
Molecular Formula: C14H18ClNO4
Formula Weight: 299.75
Storage Temperature: 2°C to 8°C
Melting Point: 179-180° dec.
MDL No.: MFCD02090708
Formula:
C₁₄H₁₈ClNO₄ MW: 299,75 g/mol Melting Pt: 179…180 °C (dec.) |
MDL Number:
MFCD02090708 CAS Number: 284493-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...