3-(Boc-amino)-3-(2-fluorophenyl)propionic acid ≥98%
Supplier: Thermo Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-3-(2-fluorophenyl)propanoic acid
L19804.03
L19804.MD
L19804.03EA
164
EUR
L19804.03
L19804.MD
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid ≥98%
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid
Purity: 98+%
CAS No.: 284493-56-7
Molecular Formula: C14H18FNO4
Formula Weight: 283.30
Storage Temperature: 2°C to 8°C
Melting Point: 142-144°
MDL No.: MFCD02090698
Formula:
C₁₄H₁₈FNO₄ MW: 283,30 g/mol Melting Pt: 142…144 °C |
MDL Number:
MFCD02090698 CAS Number: 284493-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...