3-(Boc-amino)-3-(3-fluorophenyl)propionic acid 99%
Supplier: Thermo Scientific
Synonyms:
3-[(tert-Butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid, 3-((tert-Butoxycarbonyl)amino)-3-(3-fluorophenyl)propanoic acid
L19805.03
L19805.MD
L19805.03EA
72.7
EUR
L19805.03
L19805.MD
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid 99%
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid
Purity: 99%
CAS No.: 284493-59-0
Molecular Formula: C14H18FNO4
Formula Weight: 283.30
Melting Point: 124-126°
MDL No.: MFCD02090701
Formula:
C₁₄H₁₈FNO₄ MW: 283,30 g/mol Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD02090701 CAS Number: 284493-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...