3-(Boc-amino)-3-(4-fluorophenyl)propionic acid ≥98%
Supplier: Thermo Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid, 3-[(tert-Butoxycarbonyl)amino]-3-(4-fluorophenyl)propanoic acid
L19806.03
L19806.MD
L19806.03EA
168
EUR
L19806.03
L19806.MD
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid ≥98%
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid
Purity: 98+%
CAS No.: 284493-72-7
Molecular Formula: C14H18FNO4
Formula Weight: 283.30
Storage Temperature: 2°C to 8°C
Melting Point: 152-154° dec.
MDL No.: MFCD02090715
Formula:
C₁₄H₁₈FNO₄ MW: 283,30 g/mol Melting Pt: 152…154 °C (dec.) |
MDL Number:
MFCD02090715 CAS Number: 284493-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...