N-Boc-4-bromoaniline ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Bromoaniline, N-BOC protected, tert-Butyl (4-bromophenyl)carbamate

H55226.03 H55226.06
H55226.03EA 21 EUR
H55226.03 H55226.06
N-Boc-4-bromoaniline ≥97%
N-Boc-4-bromoaniline
Formula: C₁₁H₁₄BrNO₂
MW: 272,14 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01006612
CAS Number: 131818-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR