4-(Boc-amino)benzonitrile 97%

Supplier: Thermo Fisher Scientific
Danger

H54272.MD H54272.03 H54272.06
H54272.MDEA 0 EUR
H54272.MD H54272.03 H54272.06
4-(Boc-amino)benzonitrile 97%
4-(Boc-amino)benzonitrile

Purity: 97% CAS No.: 143090-18-0 Molecular Formula: C12H14N2O2 Formula Weight: 218.26 Beilstein Reference: 6326835 MDL No.: MFCD08703141

Formula: C₁₂H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08703141
CAS Number: 143090-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR