4-Aminocarbonyl-3-nitrobenzoic acid 98%

Supplier: Thermo Scientific
Warning

B25178.03
B25178.03EA 31.5 EUR
B25178.03
4-Aminocarbonyl-3-nitrobenzoic acid 98%
4-Aminocarbonyl-3-nitrobenzoic acid

Purity: 98% CAS No.: 87594-59-0 Molecular Formula: C8H6N2O5 Formula Weight: 210.15 MDL No.: MFCD04039778

Formula: C₈H₆N₂O₅
MW: 210,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039778
CAS Number: 87594-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR