4-(Methylsulphonyl)aniline hydrochloride ≥99%
Supplier: Thermo Scientific
|
Danger
|
B23202.06
B23202.14
B23202.22
B23202.06EA
45.8
EUR
B23202.06
B23202.14
B23202.22
4-(Methylsulphonyl)aniline hydrochloride ≥99%
4-(Methylsulphonyl)aniline hydrochloride
Purity: 99%
CAS No.: 177662-76-9
Molecular Formula: C7H9NO2S.HCl
Formula Weight: 207.67
Melting Point: 237-239°
MDL No.: MFCD00216490
Formula:
C₇H₁₀ClNO₂S MW: 207,67 g/mol Melting Pt: 237…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216490 CAS Number: 177662-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...