4-Cyanophenyl-4-hexylbenzoate ≥99%
Supplier: Thermo Scientific
|
Warning
|
B22471.03
B22471.06
B22471.03EA
59.2
EUR
B22471.03
B22471.06
4-Cyanophenyl-4-hexylbenzoate ≥99%
4-Cyanophenyl 4-hexylbenzoate
Purity: 99%
CAS No.: 50793-85-6
Molecular Formula: C20H21NO2
Formula Weight: 307.39
Melting Point: 44-47°
MDL No.: MFCD00600506
Formula:
C₂₀H₂₁NO₂ MW: 307,39 g/mol Melting Pt: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00600506 CAS Number: 50793-85-6 EINECS: 256-767-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...