N-Methyl-3,4,5-trimethoxybenzylamine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Methyl-1-(3,4,5-trimethoxyphenyl)methanamine

B20996.06 B20996.14
B20996.06EA 0 EUR
B20996.06 B20996.14
N-Methyl-3,4,5-trimethoxybenzylamine 97%
N-Methyl-3,4,5-trimethoxybenzylamine

Purity: 97% CAS No.: 58780-82-8 Molecular Formula: C11H17NO3 Formula Weight: 211.26 MDL No.: MFCD00798597

Formula: C₁₁H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00798597
CAS Number: 58780-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR