5-Nitro-2-picoline 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Methyl-5-nitropyridine

H61750.06 H61750.14
H61750.06EA 79.8 EUR
H61750.06 H61750.14
5-Nitro-2-picoline 95%
5-Nitro-2-picoline
Formula: C₆H₆N₂O₂
MW: 138.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114179
CAS Number: 21203-68-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR