4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester

Supplier: Thermo Scientific

Synonyms: 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester, Piperidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

H51111.03 H51111.06 H51111.14
H51111.03EA 46.5 EUR
H51111.03 H51111.06 H51111.14
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester

CAS No.: 938043-31-3 Molecular Formula: C18H26BNO3 Formula Weight: 315.21 MDL No.: MFCD09027083

Formula: C₁₈H₂₆BNO₃
MW: 315,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027083
CAS Number: 938043-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR