3'-Bromo-2'-fluoroacetophenone 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(3-Bromo-2-fluorophenyl)ethanone

H62708.MD H62708.03
H62708.MDEA 0 EUR
H62708.MD H62708.03
3'-Bromo-2'-fluoroacetophenone 96%
3'-Bromo-2'-fluoroacetophenone

Purity: 96% CAS No.: 161957-61-5 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD09264507
CAS Number: 161957-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR