Citronellic acid ≥94%

Supplier: Thermo Scientific
Warning

B22862.14 B22862.22 B22862.36
B22862.14EA 55.3 EUR
B22862.14 B22862.22 B22862.36
Citronellic acid ≥94%
Citronellic acid

Purity: 94% CAS No.: 502-47-6 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 110°(230°F) Refractive Index: 1.4535 Boiling Point: 121-122°/1mm UN No.: 2810 MDL No.: MFCD00002728

Formula: C₁₀H₁₈O₂
MW: 170,25 g/mol
Boiling Pt: 121…122 °C (1 mmHg)
Density: 0,923
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002728
CAS Number: 502-47-6
EINECS: 207-939-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR