1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers) ≥94%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Tri-n-butyl(1-propenyl)tin
L20200.03
L20200.06
L20200.03EA
149
EUR
L20200.03
L20200.06
1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers) ≥94%
1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers)
Purity: 94%
CAS No.: 105494-65-3
Molecular Formula: C15H32Sn
Formula Weight: 331.13
Storage Temperature: 2°C to 8°C
UN No.: 2788
MDL No.: MFCD03425868
Formula:
C₁₅H₃₂Sn MW: 331,13 g/mol Boiling Pt: 77…78 °C (0,25 mmHg) |
MDL Number:
MFCD03425868 CAS Number: 105494-65-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...