(3-Methyl-2-butenyl)tributyltin 95%

Supplier: Thermo Scientific
Danger

Synonyms: Tri-n-butyl(3-methyl-2-butenyl)tin

L20214.03 L20214.06
L20214.03EA 71.8 EUR
L20214.03 L20214.06
(3-Methyl-2-butenyl)tributyltin 95%
(3-Methyl-2-butenyl)tributyltin

Purity: 95% CAS No.: 53911-92-5 Molecular Formula: C17H36Sn Formula Weight: 359.19 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4880 Boiling Point: 283° Beilstein Reference: 3588742 UN No.: 2788 MDL No.: MFCD01863649

Formula: C₁₇H₃₆Sn
MW: 359,19 g/mol
Boiling Pt: 283 °C
Density: 1,042
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD01863649
CAS Number: 53911-92-5
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR