N,N-Dioctylaniline ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: N,N-Di-n-octylaniline

H26644.03 H26644.06
H26644.03EA 60.3 EUR
H26644.03 H26644.06
N,N-Dioctylaniline ≥98%
N,N-Dioctylaniline

Purity: 98% CAS No.: 3007-75-8 Molecular Formula: C22H39N Formula Weight: 317.56 Refractive Index: 1.5495

Formula: C₂₂H₃₉N
MW: 317,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00155305
CAS Number: 3007-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR