2-Methoxy-5-isopropylbenzaldehyde ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Isopropyl-2-methoxybenzaldehyde
L14737.14
L14737.06
L14737.22
L14737.14EA
176
EUR
L14737.14
L14737.06
L14737.22
2-Methoxy-5-isopropylbenzaldehyde ≥97%
2-Methoxy-5-isopropylbenzaldehyde
Purity: 97%
CAS No.: 85902-68-7
Molecular Formula: C11H14O2
Formula Weight: 178.23
Refractive Index: 1.5390
Boiling Point: 96°/1mm
MDL No.: MFCD03095324
Formula:
C₁₁H₁₄O₂ MW: 178,23 g/mol Boiling Pt: 96 °C (1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD03095324 CAS Number: 85902-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...