4-Decylbiphenyl ≥98%

Supplier: Thermo Fisher Scientific

A16685.03 A16685.06
A16685.03EA 0 EUR
A16685.03 A16685.06
4-Decylbiphenyl ≥98%
4-Decylbiphenyl

Purity: 98% CAS No.: 93972-02-2 Molecular Formula: C22H30 Formula Weight: 294.48 Melting Point: 52-54° Boiling Point: 204-210°/1.5mm MDL No.: MFCD00102117

Formula: C₂₂H₃₀
MW: 294.48 g/mol
Boiling Pt: 204…210 °C (1.5 mmHg)
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00102117
CAS Number: 93972-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR