4-Butylbenzylamine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
B22316.06
B22316.14
B22316.06EA
106
EUR
B22316.06
B22316.14
4-Butylbenzylamine 98%
4-Butylbenzylamine
Purity: 98%
CAS No.: 57802-79-6
Molecular Formula: C11H17N
Formula Weight: 163.26
Boiling Point: 89-91°/1mm
UN No.: 2735
MDL No.: MFCD02258855
Formula:
C₁₁H₁₇N Boiling Pt: 89…91 °C (1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD02258855 CAS Number: 57802-79-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...