2-(4-Methylphenyl)ethanol ≥99%

Supplier: Thermo Scientific

Synonyms: 2-(p-Tolyl)ethanol

B20918.06 B20918.14
B20918.06EA 89 EUR
B20918.06 B20918.14
2-(4-Methylphenyl)ethanol ≥99%
2-(4-Methylphenyl)ethanol

Purity: 99% CAS No.: 699-02-5 Molecular Formula: C9H12O Formula Weight: 136.20 Flash Point: 107°(225°F) Refractive Index: 1.5260 Boiling Point: 244-245° MDL No.: MFCD00002904

Formula: C₉H₁₂O
MW: 136,20 g/mol
Boiling Pt: 244…245 °C
Density: 0,980
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002904
CAS Number: 699-02-5
EINECS: 211-824-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR