Diethoxymethylsilane 96%

Supplier: Thermo Scientific
Danger

Synonyms: Diethoxy(methyl)silane

A10153.06 A10153.09 A10153.18
A10153.06EA 44.1 EUR
A10153.06 A10153.09 A10153.18
Diethoxymethylsilane 96%
Diethoxymethylsilane

Purity: 96% CAS No.: 2031-62-1 Molecular Formula: C5H14O2Si Formula Weight: 134.25 Flash Point: 10°(50°F) Refractive Index: 1.3760 Boiling Point: 94-95° Beilstein Reference: 1733975 UN No.: 1993 MDL No.: MFCD00026749

Formula: C₅H₁₄O₂Si
MW: 134,25 g/mol
Boiling Pt: 94…95 °C
Density: 0,840
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026749
CAS Number: 2031-62-1
EINECS: 217-982-3
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR