1-Aminopiperidine 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L14078.06
L14078.14
L14078.22
L14078.06EA
60.9
EUR
L14078.06
L14078.14
L14078.22
1-Aminopiperidine 97%
1-Aminopiperidine
Purity: 97%
CAS No.: 2213-43-6
Molecular Formula: C5H12N2
Formula Weight: 100.16
Flash Point: 36°(96°F)
Refractive Index: 1.4750
Boiling Point: 145-147°
Beilstein Reference: 383565
UN No.: 2733
MDL No.: MFCD00006489
Formula:
C₅H₁₂N₂ Boiling Pt: 145…147 °C Density: 0,928 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006489 CAS Number: 2213-43-6 EINECS: 218-666-8 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...