Methyl-4-piperidinecarboxylate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 4-Piperidinecarboxylate, Methyl isonipecotate, Methyl piperidine-4-carboxylate
H61754.14
H61754.22
H61754.14EA
55.9
EUR
H61754.14
H61754.22
Methyl-4-piperidinecarboxylate ≥98%
Methyl 4-piperidinecarboxylate
Formula:
C₇H₁₃NO₂ MW: 143,18 g/mol Boiling Pt: 85…90 °C Density: 1,06 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190578 CAS Number: 2971-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...