4-Bromo-o-anisidine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Bromo-2-methoxyaniline, 2-Amino-5-bromoanisole, 2-Methoxy-4-bromoaniline
H61454.03
H61454.06
H61454.03EA
0
EUR
H61454.03
H61454.06
4-Bromo-o-anisidine ≥98%
4-Bromo-o-anisidine
Purity: 98%
CAS No.: 59557-91-4
Molecular Formula: C7H8BrNO
Formula Weight: 202.05
UN No.: 2811
MDL No.: MFCD01204266
Formula:
C₇H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD01204266 CAS Number: 59557-91-4 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...