4-Fluoro-2-nitrobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H61971.03
H61971.06
H61971.03EA
54.3
EUR
H61971.03
H61971.06
4-Fluoro-2-nitrobenzaldehyde ≥98%
4-Fluoro-2-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01666439 CAS Number: 2923-96-8 EINECS: 220-884-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...