2-Amino-3-nitro-5-fluoropyridine 98%

Supplier: Thermo Fisher Scientific
Warning

H61293.03 H61293.06
H61293.03EA 0 EUR
H61293.03 H61293.06
2-Amino-3-nitro-5-fluoropyridine 98%
2-Amino-3-nitro-5-fluoropyridine
Formula: C₅H₄FN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04972413
CAS Number: 212268-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR