1,14-Tetradecanedioic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H60254.06 H60254.14
H60254.06EA 64.4 EUR
H60254.06 H60254.14
1,14-Tetradecanedioic acid ≥98%
1,14-Tetradecanedioic acid
Formula: C₁₄H₂₆O₄
Boiling Pt: 407 °C (1013 hPa)
Melting Pt: 124…127 °C
Density: 1,108 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002743
CAS Number: 821-38-5
EINECS: 212-476-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR