3-Bromo-5-fluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

H61544.14 H61544.22
H61544.14EA 106 EUR
H61544.14 H61544.22
3-Bromo-5-fluorotoluene ≥98%
3-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
Storage Temperature: Ambient
MDL Number: MFCD01861195
CAS Number: 202865-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR