Ethyl fluoroacetate 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Fluoroacetic ethyl ester

H61340.09 H61340.18
H61340.09EA 40.6 EUR
H61340.09 H61340.18
Ethyl fluoroacetate 98
Ethyl fluoroacetate
Formula: C₄H₇FO₂
MW: 106.1 g/mol
Boiling Pt: 119…120 °C
Density: 1.093
Storage Temperature: Ambient
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
UN: 2929
ADR: 6.1 II
Merck Index: 14,4167

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR