4-Aminobenzhydrazide

Supplier: Cayman Chemical

Synonyms: 4-Aminobenzoic hydrazide, 4-Aminobenzene-1-carbohydrazide, 4-Aminobenzoic acid hydrazide, 4-Aminobenzohydrazide

14845-500 14845-1
CAYM14845-500EA 62.3 EUR
CAYM14845-500 CAYM14845-1
4-Aminobenzhydrazide
4-Aminobenzhydrazide
Formula: C₇H₉N₃O
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 280…283 °C
MDL Number: MFCD00007606
CAS Number: 5351-17-7
EINECS: 226-324-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR