1-Fluoro-2-ethynylbenzene ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene
H60596.MD
H60596.03
H60596.MDEA
49.2
EUR
H60596.MD
H60596.03
1-Fluoro-2-ethynylbenzene ≥97%
1-Fluoro-2-ethynylbenzene
Formula:
C₈H₅F MW: 120,13 g/mol Boiling Pt: 150 °C Density: 1,060 Flash Pt: 37 °C (99 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00799540 CAS Number: 766-49-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...