1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Methylbis(trimethylsilyloxy)silane

H60586.AD H60586.AK
H60586.ADEA 146 EUR
H60586.AD H60586.AK
1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥97%
1,1,1,3,5,5,5-Heptamethyltrisiloxane
Formula: C₇H₂₂O₂Si₃
MW: 222,51 g/mol
Boiling Pt: 142 °C
Density: 0,819
Flash Pt: 22 °C (72 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048000
CAS Number: 1873-88-7
EINECS: 217-496-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR