3-Bromo-2-nitroaniline ≥98%
Supplier: Thermo Scientific
|
Warning
|
H61169.03
H61169.06
H61169.03EA
98.1
EUR
H61169.03
H61169.06
3-Bromo-2-nitroaniline ≥98%
3-Bromo-2-nitroaniline
Purity: 98%
CAS No.: 7138-15-0
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Physical Form: Crystalline
Appearance: Dark red
UN No.: 2811
MDL No.: MFCD08063248
Formula:
C₆H₅BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08063248 CAS Number: 7138-15-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...