Azacyclonol 95%

Supplier: Thermo Scientific
Warning

Synonyms: alpha,alpha-Diphenyl-4-piperidinemethanol, alpha-(4-Piperidyl)benzhydrol, Diphenyl(4-piperidinyl)methanol, α,α-Diphenyl-4-piperidinomethanol, Diphenyl-4-piperidinemethanol

H61281.14 H61281.22
H61281.14EA 0 EUR
H61281.14 H61281.22
Azacyclonol 95%
Azacyclonol
Formula: C₁₈H₂₁NO
MW: 267,37 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00066980
CAS Number: 115-46-8
EINECS: 204-092-5
Merck Index: 13,895

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR