4-Bromo-2-fluorophenylacetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H61176.06 H61176.14
H61176.06EA 131 EUR
H61176.06 H61176.14
4-Bromo-2-fluorophenylacetic acid ≥98%
4-Bromo-2-fluorophenylacetic acid
Formula: C₈H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09032952
CAS Number: 114897-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR