Funnels, powder
Supplier: SMITH SCIENTIFIC LTD
2356-80 2356-100 2356-120
SMIT2356-80EA 0 EUR
SMIT2356-80 SMIT2356-100 SMIT2356-120
Funnels, powder
Funnels Powder Funnels
Borosilicate glass.
  • Good chemical and physical properties
  • Excellent thermal resistance
  • Quick and easy to clean
With smooth internal walls. For transferring powders. Can also be used for liquids.