Glass tubing, capillary
Supplier: SMITH SCIENTIFIC LTD
TC/10/3.5 TC/8/3.0 TC/10/3.0 TTC/10/2.0
TC/10/3.5EA 171170 EUR
TC/10/3.5 SMITTC/8/3.0 SMITTC/10/3.0 SMITTTC/10/2.0
Glass tubing, capillary
Tubing Transfer Tubing
Made from Simax glass.
  • Smooth, non-porous, transparent
  • Catalytically indifferent
  • Corrosion resistant (even in exacting operations)
  • Sufficiently homogenous and without presence of heterogenous particles
Capillary tubings are used in such applications as the production of laboratory glassware, industrial apparatus, pilot plant, pipe lines, drainage, decorative glass, solar collectors and light fittings.