Cleanroom Chairs
Supplier: TRACT
TC311-BLA TC311-BLU
TRACTC311-BLAEA 550 EUR
TRACTC311-BLA TRACTC311-BLU
Cleanroom Chairs
Chairs
These cleanroom chairs has ergonomic adjustments such as pneumatic height lift, independent seat and back tilt, lumbar and pelvic support and a spring pivot for permanent back support.
  • Stable and robust design
  • Easy clean - resistant to aggressive cleaning agents
  • Ideal for use with Laminar Flow and Isolators
  • Helps avoid lower back strain and repetitive strain injuries
  • HEPA filter fitted to seat base
Seat height range: 540 to 800 mm.
Certifications: Certified to US Federal Standard 209E.