Acid and chemical storage cabinets
Supplier: SAFETY UNLIMITED
AC2010 AC2040 AC2060 AC2070 AC2020 AC2030 AC2050 AC2080
SAFUAC2010EA 637 EUR
SAFUAC2010 SAFUAC2040 SAFUAC2060 SAFUAC2070 SAFUAC2020 SAFUAC2030 SAFUAC2050 SAFUAC2080
Acid and chemical storage cabinets
Furniture Storage Cabinets
Cabinets are constructed from Zintec coated steel with epoxy powder coat.
  • Doors have a reinforced vertical panel to prevent distortion due to heat
  • Lockable via a two point locking system
  • Fully adjustable shelves to 45 mm pitch
  • Spillages to drain into the removable leakproof sump below
  • Acid and corrosive resistance
Certifications: All cabinets comply with HSG51, COSHH and current Health, safety and environmental regulations.