D-(-)-Leucine
Supplier: G Biosciences
Synonyms:
H-D-Leu-OH, (R)-(-)-Leucine
RC-065
RC-064
RC-065EA
302
EUR
RC-065
RC-064
D-(-)-Leucine
D(-)-Leucine
Formula:
C₆H₁₃NO₂ MW: 131,17 g/mol Boiling Pt: 226 °C (1013 hPa) |
MDL Number:
MFCD00063088 CAS Number: 328-38-1 EINECS: 206-327-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...