Anti-HOXB7 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP37068_P050 ARP37068_T100
AVIVARP37068_P050EA 376 EUR
AVIVARP37068_P050 AVIVARP37068_T100
Anti-HOXB7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HOXB7 (homeobox B7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR