Anti-GEMIN2 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB1919-100 ORB1919-200
BIRBORB1919-100EA 0 EUR
BIRBORB1919-100 BIRBORB1919-200
Anti-GEMIN2 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: GEMIN2 (gem (nuclear organelle) associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR