Anti-FAM83F Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2952-100 ORB2952-200
BIRBORB2952-100EA 0 EUR
BIRBORB2952-100 BIRBORB2952-200
Anti-FAM83F Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: FAM83F (family with sequence similarity 83, member F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR