Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB12107-100 ORB12107-50
BIRBORB12107-100EA 0 EUR
BIRBORB12107-100 BIRBORB12107-50
Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
FACS: 1:50-100

Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR