Anti-CSNK2A1 Mouse monoclonal antibody

Supplier: Biorbyt
ORB22763-100 ORB22763-20 ORB22763-5
BIRBORB22763-100EA 0 EUR
BIRBORB22763-100 BIRBORB22763-20 BIRBORB22763-5
Anti-CSNK2A1 Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: CSNK2A1 (casein kinase 2, alpha 1 polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR