Anti-CHGA Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22810-100 ORB22810-20 ORB22810-5
BIRBORB22810-100EA 0 EUR
BIRBORB22810-100 BIRBORB22810-20 BIRBORB22810-5
Anti-CHGA Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CHGA (chromogranin A (parathyroid secretory protein 1))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR