Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB22924-1 ORB22924-500
BIRBORB22924-1EA 0 EUR
BIRBORB22924-1 BIRBORB22924-500
Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR