Anti-Kappa (L chain) Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100072-100 ORB100072-200
BIRBORB100072-100EA 0 EUR
BIRBORB100072-100 BIRBORB100072-200
Anti-Kappa (L chain) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Kappa (L chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR